Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam219b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166364.1 | NP_001159836.1 | 1, 2 | 300 | ||
NM_175273.4 | NP_780482.1 | 1, 2 | 300 | |||
GenBank mRNA | AK028365.1 | - | 1, 2 | 301 | ||
BC111883.1 | - | 1, 2 | 265 | 40060305 | ||
BC112403.1 | - | 1, 2 | 265 | 40060304 |