Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083891.1 | NP_001077360.1 | 2 | 211 | ||
NM_030087.2 | NP_084363.2 | 2 | 211 | |||
GenBank mRNA | AK018779.1 | - | 2 | 176 | ||
AK077433.1 | - | 2 | 152 | |||
AK154191.1 | - | 2 | 144 | |||
AK154477.1 | - | 2 | 190 | |||
AK160289.1 | - | 2 | 142 | |||
BC013640.1 | - | 2 | 168 | 4188234 | ||
BU555486.1 | - | 2 | 214 | |||
BY064612.1 | - | 2 | 179 |