Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc30a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030188.3 | NP_084464.3 | 1 | 2037 | ||
GenBank mRNA | AK019670.1 | - | 1 | 1911 | ||
AK029535.1 | - | 1840 | ||||
AK045096.1 | - | 1 | 1837 | |||
BC004773.1 | - | 1 | 1832 | 3256753 |