Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl9l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030256.2 | NP_084532.2 | 7 | 2989 | ||
GenBank mRNA | AB128033.1 | - | 6 | 2775 | ||
AY296058.1 | - | 7 | 2989 | |||
BC082304.1 | - | 6 | 3123 | 6856460 | ||
BC119466.2 | - | 2 | 523 | |||
BC119467.2 | - | 2 | 515 |