Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rilpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030259.1 | NP_084535.1 | 1, 2 | 459 | ||
GenBank mRNA | AK133760.1 | - | 1, 2 | 591 | ||
AK150123.1 | - | 1, 2 | 563 | |||
AK150252.1 | - | 1, 2 | 562 | |||
BC003324.1 | - | 1, 2 | 459 | 3156139 |