Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031185.3 | NP_112462.1 | 3 | 2228 | ||
GenBank mRNA | AB020886.1 | - | 3 | 2177 | ||
AB025278.1 | - | 3 | 2191 | |||
AB051563.1 | - | 3 | 2179 | |||
AB070852.1 | - | 2 | 1913 | |||
AB070853.1 | - | 2 | 1899 | |||
AF326228.1 | - | 3 | 2177 | |||
AF326230.1 | - | 2 | 1882 | |||
BC042461.1 | - | 1 | 1624 | |||
BC043939.1 | - | 1 | 1624 | |||
BC071185.1 | - | 3 | 2177 | |||
BC138364.1 | - | 3 | 2173 | 8861384 | ||
BC138365.1 | - | 3 | 2172 | 8861385 |