Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr137b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031999.2 | NP_114388.2 | 1 | 400 | ||
GenBank mRNA | AK050369.1 | - | 1 | 341 | ||
AK080884.1 | - | 1 | 353 | |||
AK128965.1 | - | 1 | 400 | |||
AK147921.1 | - | 1 | 372 | |||
AK172286.1 | - | 1 | 345 | |||
AK190995.1 | - | 1 | 95 | |||
AK195847.1 | - | 1 | 177 | |||
AK198577.1 | - | 1 | 95 | |||
AK204136.1 | - | 1 | 95 | |||
AK204647.1 | - | 1 | 95 | |||
BC003212.1 | - | 1 | 393 | 3157256 |
Gene Symbol: Gpr137b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF154337.1 | - | 1 | 363 | ||
AK009736.1 | - | 1 | 365 | |||
AK010724.1 | - | 1 | 378 | |||
AK087190.1 | - | 1 | 373 | |||
AK170103.1 | - | 1 | 373 | |||
AK170650.1 | - | 1 | 281 | |||
AK171335.1 | - | 1 | 373 | |||
BC093499.1 | - | 1 | 360 |