Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013580.3 | NP_001013598.1 | 2 | 192 | ||
NM_001013581.2 | NP_001013599.1 | 2 | 232 | |||
NM_001122850.1 | NP_001116322.1 | 4 | 741 | |||
NM_033620.2 | NP_296369.2 | 4 | 741 | |||
GenBank mRNA | AK052416.1 | - | 4 | 728 | ||
AK053563.1 | - | 4 | 722 | |||
AK053576.1 | - | 4 | 699 | |||
AK053762.1 | - | 4 | 727 | |||
AK137690.1 | - | 4 | 700 | |||
AY026057.1 | - | 4 | 552 | |||
AY856081.1 | - | 2 | 236 | |||
AY856082.1 | - | 2 | 232 | |||
BC090616.1 | - | 4 | 741 | 30943099 | ||
BC141187.1 | - | 4 | 503 | 9055799 | ||
BC145508.1 | - | 4 | 503 | 9054071 | ||
CF172228.1 | - | 2 | 192 |