Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053124.2 | NP_444354.2 | 18 | 2705 | ||
GenBank mRNA | AF325921.1 | - | 18 | 2357 | ||
AF375046.1 | - | 18 | 2424 | |||
AK052320.1 | - | 6 | 658 | |||
AK153789.1 | - | 18 | 3023 | |||
AK156915.1 | - | 2 | 169 | |||
AK157142.1 | - | 2716 | ||||
BC021922.1 | - | 6 | 643 | |||
BC053069.1 | - | 18 | 2705 | 6409140 |