Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025772.4 | NP_080048.2 | 6, 7 | 620 | ||
GenBank mRNA | AJ404859.1 | - | 6, 7 | 619 | ||
AK010924.1 | - | 2, 3 | 232 | |||
AK017400.1 | - | 2, 3 | 398 | |||
AK150306.1 | - | 6, 7 | 616 | |||
AK159656.1 | - | 6, 7 | 745 | |||
AY265461.1 | - | 2, 3 | 398 | |||
BC018350.1 | - | 6, 7 | 611 | 4195234 | ||
BC048682.1 | - | 6, 7 | 665 | |||
BC058574.1 | - | 6, 7 | 593 | 6487720 |