Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025772.4 | NP_080048.2 | 8, 9 | 798 | ||
GenBank mRNA | AJ404859.1 | - | 8, 9 | 797 | ||
AK010924.1 | - | 4, 5 | 410 | |||
AK017400.1 | - | 4, 5 | 576 | |||
AK159656.1 | - | 8, 9 | 923 | |||
AK180780.1 | - | 1, 2 | 92 | |||
AK189627.1 | - | 1, 2 | 92 | |||
AY265461.1 | - | 4, 5 | 576 | |||
BC018350.1 | - | 8, 9 | 789 | 4195234 | ||
BC048682.1 | - | 8, 9 | 843 | |||
BC058574.1 | - | 8, 9 | 771 | 6487720 |