Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tprn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175286.4 | NP_780495.2 | 1 | 991 | ||
GenBank mRNA | AK082888.1 | - | 1 | 972 | ||
AK165772.1 | - | 1 | 933 | |||
AK171799.1 | - | 1 | 1005 | |||
BC089508.1 | - | 1 | 501 | 6589951 |