Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028889.2 | NP_083165.1 | 2 | 416 | ||
GenBank mRNA | AK016491.1 | - | 2 | 416 | ||
AK045911.1 | - | 2 | 418 | |||
AK144175.1 | - | 2 | 419 | |||
AK171846.1 | - | 2 | 418 | |||
BC085088.1 | - | 2 | 401 | 6467625 |