Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033194.2 | NP_001028366.1 | 15 | 2193 | ||
GenBank mRNA | AK145040.1 | - | 2197 | |||
AK160997.1 | - | 15 | 2181 | |||
AK165830.1 | - | 15 | 2194 | |||
BC141157.1 | - | 15 | 2183 | 9055769 |