Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033194.2 | NP_001028366.1 | 7 | 968 | ||
GenBank mRNA | AK145040.1 | - | 970 | |||
AK160997.1 | - | 7 | 956 | |||
AK165830.1 | - | 7 | 969 | |||
AK179332.1 | - | 2 | 62 | |||
BC141157.1 | - | 7 | 958 | 9055769 |