Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240075.2 | XP_006240137.1 | 9 | 1505 | ||
XM_006240076.2 | XP_006240138.1 | 9 | 1474 | |||
XM_006240077.2 | XP_006240139.1 | 8 | 1352 | |||
GenBank mRNA | AY026526.1 | - | 8 | 1291 | ||
DQ473607.1 | - | 8 | 1246 | |||
FQ222839.1 | - | 4 | 511 |