Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240075.2 | XP_006240137.1 | 15 | 2277 | ||
XM_006240076.2 | XP_006240138.1 | 15 | 2246 | |||
XM_006240077.2 | XP_006240139.1 | 14 | 2124 | |||
GenBank mRNA | FQ222839.1 | - | 10 | 1283 |