Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276714.1 | NP_001263643.1 | 6 | 1559 | ||
XM_006235296.2 | XP_006235358.1 | 8 | 1896 | |||
XM_006235297.1 | XP_006235359.1 | 8 | 1793 | |||
XM_008762295.1 | XP_008760517.1 | 9 | 1965 | |||
XM_008762296.1 | XP_008760518.1 | 8 | 1793 | |||
XM_008762297.1 | XP_008760519.1 | 7, 8 | 1760 | |||
XM_008762299.1 | XP_008760521.1 | 8 | 1793 |