Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057213.2 | NP_476561.1 | 11, 12 | 1165 | ||
GenBank mRNA | BC085714.1 | - | 11, 12 | 1165 | 7109340 | |
Y12635.1 | - | 11, 12 | 1166 |