Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080688.1 | NP_542419.1 | 15 | 2233 | ||
XM_006245135.2 | XP_006245197.1 | 17 | 3404 | |||
XM_006245136.2 | XP_006245198.1 | 16 | 2245 | |||
XM_006245137.2 | XP_006245199.1 | 16 | 2544 | |||
XM_006245138.2 | XP_006245200.1 | 16 | 2419 | |||
XM_008767182.1 | XP_008765404.1 | 16 | 2823 | |||
XM_008767183.1 | XP_008765405.1 | 16 | 2823 | |||
XM_008767184.1 | XP_008765406.1 | 16 | 2862 | |||
XM_008767185.1 | XP_008765407.1 | 16 | 2246 | |||
XM_008767186.1 | XP_008765408.1 | 16 | 2824 | |||
XM_008767187.1 | XP_008765409.1 | 16 | 2821 | |||
XM_008767188.1 | XP_008765410.1 | 16 | 2769 | |||
XM_008767189.1 | XP_008765411.1 | 16 | 2823 | |||
XM_008767190.1 | XP_008765412.1 | 15 | 2726 | |||
XM_008767191.1 | XP_008765413.1 | 15 | 2723 | |||
GenBank mRNA | D50455.1 | - | 15 | 2223 | ||
U16655.1 | - | 15 | 2233 |