Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080689.4 | NP_542420.1 | 5 | 661 | ||
GenBank mRNA | X54531.1 | - | 5 | 661 |