Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130411.2 | NP_569095.1 | 3 | 409 | ||
XM_006230204.2 | XP_006230266.1 | 3 | 493 | |||
GenBank mRNA | AF416730.1 | - | 2 | 305 | ||
AF495469.1 | - | 3 | 409 | |||
BC086971.1 | - | 3 | 385 | 7124429 | ||
FQ226379.1 | - | 3 | 397 | |||
FQ226941.1 | - | 3 | 429 | |||
FQ233306.1 | - | 3 | 403 | |||
FQ233390.1 | - | 3 | 405 | |||
FQ233414.1 | - | 3 | 407 | |||
FQ233980.1 | - | 3 | 322 | |||
FQ234750.1 | - | 3 | 407 |