Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130411.2 | NP_569095.1 | 9 | 1134 | ||
XM_006230204.2 | XP_006230266.1 | 9 | 1218 | |||
GenBank mRNA | AF416730.1 | - | 8 | 1030 | ||
AF495469.1 | - | 9 | 1134 | |||
BC086971.1 | - | 9 | 1110 | 7124429 | ||
FQ225277.1 | - | 6 | 656 | |||
FQ225360.1 | - | 6 | 656 | |||
FQ225522.1 | - | 5 | 573 | |||
FQ226290.1 | - | 7 | 603 | |||
FQ226379.1 | - | 9 | 1125 | |||
FQ226941.1 | - | 9 | 1154 | |||
FQ233306.1 | - | 9 | 1128 | |||
FQ233390.1 | - | 9 | 1130 | |||
FQ233414.1 | - | 9 | 1132 | |||
FQ233980.1 | - | 9 | 1048 | |||
FQ234750.1 | - | 9 | 1136 |