Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130426.4 | NP_569110.1 | 8, 9 | 979 | ||
GenBank mRNA | AF142499.1 | - | 4, 5 | 466 | ||
AF498039.2 | - | 8, 9 | 979 | |||
AY191268.1 | - | 979 | ||||
AY191269.1 | - | 979 | ||||
AY344841.1 | - | 8, 9 | 892 |