Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133531.2 | NP_598215.1 | 7 | 1018 | ||
GenBank mRNA | AF399873.1 | - | 7 | 1018 | ||
FQ223969.1 | - | 7 | 1025 | |||
FQ224186.1 | - | 7 | 1024 |