Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182842.1 | NP_878262.1 | 3 | 227 | ||
XM_006250470.2 | XP_006250532.1 | 3 | 344 | |||
GenBank mRNA | AB101231.1 | - | 3 | 227 | ||
AB101232.1 | - | 3 | 227 | |||
D86557.1 | - | 3 | 233 |