Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Spata7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138862.3 | NP_620217.2 | 6 | 644 | ||
XM_006240413.2 | XP_006240475.1 | 5 | 627 | |||
XM_006240414.2 | XP_006240476.1 | 6 | 719 | |||
XM_006240415.2 | XP_006240477.1 | 4 | 552 | |||
XM_008764737.1 | XP_008762959.1 | 5 | 598 | |||
GenBank mRNA | CK470849.1 | - | 2 | 308 | ||
CK600391.1 | - | 5 | 616 |