Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138862.3 | NP_620217.2 | 6 | 852 | ||
XM_006240413.2 | XP_006240475.1 | 5 | 835 | |||
XM_006240414.2 | XP_006240476.1 | 6 | 927 | |||
XM_006240415.2 | XP_006240477.1 | 4 | 760 | |||
XM_008764737.1 | XP_008762959.1 | 5 | 806 | |||
GenBank mRNA | CK470849.1 | - | 2 | 516 |