Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138891.1 | NP_620246.1 | 1 | 896 | ||
GenBank mRNA | AY030276.1 | - | 1 | 896 |
Set Membership: |
![]() |