Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcyap1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270579.1 | NP_001257508.1 | 12, 13 | 1279 | ||
NM_001270580.1 | NP_001257509.1 | 12, 13 | 1279 | |||
NM_001270581.1 | NP_001257510.1 | 13, 14 | 1391 | |||
NM_001270582.1 | NP_001257511.1 | 12, 13 | 1279 | |||
NM_001270583.1 | NP_001257512.1 | 12, 13 | 1279 | |||
NM_133511.2 | NP_598195.1 | 12, 13 | 1279 | |||
XM_008762870.1 | XP_008761092.1 | 11, 12 | 1025 | |||
GenBank mRNA | D14908.1 | - | 13, 14 | 1271 | ||
D14909.1 | - | 12, 13 | 1069 | |||
D16465.1 | - | 12, 13 | 1279 | |||
L16506.1 | - | 6, 7 | 600 | |||
L16680.1 | - | 11, 12 | 972 | |||
Z22735.1 | - | 6, 7 | 600 | |||
Z23272.1 | - | 11, 12 | 1032 | |||
Z23273.1 | - | 11, 12 | 1032 | |||
Z23274.1 | - | 11, 12 | 1032 | |||
Z23275.1 | - | 11, 12 | 1032 | |||
Z23279.1 | - | 11, 12 | 1032 | |||
Z23282.1 | - | 12, 13 | 1335 |
Set Membership: |
![]() |