Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012499.1 | NP_036631.1 | 15 | 3526 | ||
XM_006254506.2 | XP_006254568.1 | 16 | 3786 | |||
XM_006254508.2 | XP_006254570.1 | 17 | 3627 | |||
XM_006254509.2 | XP_006254571.1 | 17 | 3596 | |||
XM_006254510.2 | XP_006254572.1 | 15 | 3732 | |||
XM_006254511.2 | XP_006254573.1 | 16 | 3573 | |||
XM_006254512.2 | XP_006254574.1 | 16 | 4235 | |||
XM_006254513.2 | XP_006254575.1 | 15 | 3681 | |||
XM_006254515.2 | XP_006254577.1 | 17 | 3452 | |||
XM_006254518.2 | XP_006254580.1 | 16 | 4050 | |||
XM_006254520.2 | XP_006254582.1 | 16 | 3483 | |||
XM_006254521.2 | XP_006254583.1 | 16 | 3270 | |||
XM_008772015.1 | XP_008770237.1 | 16 | 5718 | |||
XM_008772016.1 | XP_008770238.1 | 16 | 5668 | |||
XM_008772017.1 | XP_008770239.1 | 16 | 4059 | |||
GenBank mRNA | D38629.1 | - | 15 | 3526 | ||
L19303.1 | - | 1 | 109 |