Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cftr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031506.1 | NP_113694.1 | 14 | 1971 | ||
XM_006236097.2 | XP_006236159.1 | 15 | 2090 | |||
XM_008762680.1 | XP_008760902.1 | 15 | 2090 | |||
GenBank mRNA | FJ959076.1 | - | 14 | 1729 | ||
M89906.1 | - | 4 | 391 |