Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012527.2 | NP_036659.2 | 5 | 1750 | ||
XM_008771690.1 | XP_008769912.1 | 6 | 1696 | |||
GenBank mRNA | AB017656.1 | - | 1 | 1203 | ||
M16407.1 | - | 2 | 1243 | |||
M18088.1 | - | 3 | 1334 |
Set Membership: |
![]() |