Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012572.1 | NP_036704.1 | 16 | 2103 | ||
GenBank mRNA | U08257.1 | - | 16 | 2103 | ||
X59996.1 | - | 16 | 2104 |