Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nupl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139091.2 | NP_620791.1 | 13 | 1536 | ||
XM_006252093.2 | XP_006252155.1 | 13 | 1641 | |||
GenBank mRNA | AF000900.1 | - | 12 | 1407 | ||
BC085690.1 | - | 13 | 1536 | 7190650 | ||
U63839.1 | - | 13 | 1549 |