Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cd48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139103.1 | NP_620803.1 | 3 | 509 | ||
GenBank mRNA | BC060542.1 | - | 3 | 546 | 6921213 | |
FQ228817.1 | - | 3 | 563 | |||
FQ234539.1 | - | 3 | 543 | |||
X13016.1 | - | 3 | 509 |