Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adipoq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144744.3 | NP_653345.1 | 3 | 529 | ||
GenBank mRNA | AY033885.1 | - | 3 | 476 | ||
BC092565.1 | - | 2 | 465 | 7308618 | ||
FM047633.1 | - | 3 | 529 |