Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145081.3 | NP_659549.2 | 12 | 1639 | ||
XM_006254262.2 | XP_006254324.1 | 15 | 1974 | |||
XM_006254263.2 | XP_006254325.1 | 13 | 1793 | |||
XM_006254264.2 | XP_006254326.1 | 13 | 1790 | |||
XM_008771852.1 | XP_008770074.1 | 15 | 1971 | |||
XM_008771853.1 | XP_008770075.1 | 12 | 1659 | |||
GenBank mRNA | AB050777.1 | - | 7 | 1589 | ||
AB069907.3 | - | 12 | 1639 | |||
AB222073.1 | - | 7 | 1368 | |||
AB222074.1 | - | 12 | 1590 | |||
BC086976.1 | - | 13 | 1605 | 7111026 | ||
FQ223875.1 | - | 13 | 1793 |