Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012667.2 | NP_036799.1 | 908 | |||
GenBank mRNA | J05097.1 | - | 908 | |||
M31477.1 | - | 1 | 365 |
Set Membership: |
![]() |