Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017053.1 | NP_058749.1 | 4 | 1159 | ||
GenBank mRNA | J05189.1 | - | 4 | 1159 |
Set Membership: |
G-protein coupled receptor |