Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031538.2 | NP_113726.1 | 2, 3 | 418 | ||
XM_006236630.2 | XP_006236692.1 | 3, 4 | 1047 | |||
XM_008762976.1 | XP_008761198.1 | 3, 4 | 795 | |||
GenBank mRNA | BC088126.1 | - | 2, 3 | 418 | 7381324 | |
X03015.1 | - | 2, 3 | 457 |