- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Htr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022225.1 | NP_071561.1 | 1 | 446 | ||
GenBank mRNA | X62944.1 | - | 1 | 482 |
Set Membership: |
![]() |