Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145723.1 | NP_663775.1 | 5, 6 | 652 | ||
XM_006238024.2 | XP_006238086.1 | 6, 7 | 730 | |||
GenBank mRNA | AF115435.1 | - | 5, 6 | 652 |