Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012855.2 | NP_036987.2 | 2 | 95 | ||
XM_008771064.1 | XP_008769286.1 | 2 | 1193 | |||
GenBank mRNA | D28508.1 | - | 2 | 95 |