Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adarb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111055.1 | NP_001104525.1 | 7 | 1692 | ||
NM_001111056.1 | NP_001104526.1 | 7 | 1739 | |||
NM_001111057.1 | NP_001104527.1 | 7 | 1739 | |||
NM_012894.2 | NP_037026.2 | 7 | 1692 | |||
XM_006256275.2 | XP_006256337.1 | 6 | 2242 | |||
XM_006256276.2 | XP_006256338.1 | 6 | 2242 | |||
XM_006256277.2 | XP_006256339.1 | 6 | 1596 | |||
XM_006256278.2 | XP_006256340.1 | 6 | 3836 | |||
XM_008772864.1 | XP_008771086.1 | 7 | 1652 | |||
XM_008772865.1 | XP_008771087.1 | 5 | 4497 | |||
GenBank mRNA | CB556744.1 | - | 4 | 566 | ||
U43534.1 | - | 6 | 1439 |