- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012993.2 | NP_037125.1 | 17, 18 | 2097 | ||
XM_006238499.2 | XP_006238561.1 | 19, 20 | 2435 | |||
XM_006238500.2 | XP_006238562.1 | 18, 19 | 2299 | |||
GenBank mRNA | FQ218095.1 | - | 17, 18 | 2220 | ||
L27124.1 | - | 17, 18 | 2098 | |||
X93208.1 | - | 19, 20 | 2302 |