Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oaz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297557.1 | NP_001284486.1 | 5 | 1022 | ||
NM_001301048.1 | NP_001287977.1 | 5 | 1016 | |||
NM_139081.2 | NP_620781.1 | 5 | 1022 | |||
GenBank mRNA | D10706.1 | - | 5 | 994 | ||
FQ211375.1 | - | 5 | 1022 | |||
FQ213504.1 | - | 5 | 1022 | |||
FQ213794.1 | - | 5 | 1022 | |||
FQ214019.1 | - | 5 | 1017 | |||
FQ214051.1 | - | 5 | 1017 | |||
FQ214373.1 | - | 5 | 1017 | |||
FQ214458.1 | - | 5 | 1018 | |||
FQ216817.1 | - | 5 | 1022 | |||
FQ218486.1 | - | 5 | 1017 | |||
FQ218990.1 | - | 5 | 1017 | |||
FQ220491.1 | - | 5 | 1017 | |||
FQ225116.1 | - | 5 | 1017 | |||
FQ225909.1 | - | 5 | 1017 | |||
FQ225949.1 | - | 5 | 1020 | |||
FQ230452.1 | - | 5 | 1017 | |||
FQ231088.1 | - | 5 | 1017 | |||
FQ232667.1 | - | 5 | 1017 | |||
FQ232997.1 | - | 5 | 1017 | |||
FQ233723.1 | - | 5 | 1018 |