Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013079.2 | NP_037211.2 | 3 | 605 | ||
XM_006236089.2 | XP_006236151.1 | 3 | 598 | |||
GenBank mRNA | BC081719.1 | - | 3 | 605 | 7132312 | |
U07201.1 | - | 3 | 549 | |||
U07202.1 | - | 3 | 549 |