Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100922.1 | NP_001094392.1 | 9 | 921 | ||
XM_006255219.1 | XP_006255281.1 | 9 | 855 | |||
GenBank mRNA | AY375243.1 | - | 9 | 767 | ||
BC129128.1 | - | 9 | 921 | 7314651 | ||
X57986.1 | - | 9 | 831 |