Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ass1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013157.3 | NP_037289.1 | 1 | 86 | ||
XM_006233911.2 | XP_006233973.1 | 3 | 237 | |||
GenBank mRNA | BC063146.1 | - | 3 | 227 | 6888536 | |
M36708.1 | - | 1 | 86 | |||
X12459.1 | - | 1 | 86 |